Dropping bottles, LDPE, Azlon®
Proveedor: SciLabware
BGE814PNC BGE820PNC BGE828PNC BGE830PNC
215-0482EA 23.2 EUR
215-0482 215-0483 215-0485 215-0487
Dropping bottles, LDPE, Azlon®
Frascos Frascos de goteo
LDPE, translucent.
  • Closure features a captive sealing cap and nozzle can be cut to adjust flow rate
  • Good chemical resistance
  • Heat resistant up to +80 °C
With spouted dropper closure. Suitable for use as a dropping or wash bottle.