Botellas de lavado, boca estrecha
Proveedor: KARTELL
680 681 682
215-1365EA 4.9 EUR
215-1365 215-1366 215-1367
Botellas de lavado, boca estrecha
Frascos Frascos de lavado
LDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Suitable for foodstuff
The stream of liquid is obtained by blowing through the straight tube. The direction of the stream of liquid can be controlled by hand.