Wash bottles, integral, wide neck, Azlon®
Proveedor: SciLabware
WGW800P
215-4162EA 45.1 EUR
215-4162
Wash bottles, integral, wide neck, Azlon®
Frascos Frascos de lavado
Wide neck wash bottles with red, yellow, blue, green and orange PP caps.
  • Fitted with colour coded one-piece PP closure that maintains it's shape in use
  • Designed with integral spout to prevent contamination
  • Good chemical resistance
The round iIntegral wash bottle is moulded with an integral spout so you can say goodbye to the contamination caused by the inner tube touches the benchtop during re-filling.