WHEATON® Jars, Wide Mouth, with Thread
Proveedor: DWK Life Sciences
W216922 W216923 W216903 W216904 W216905 W216906 W216907 W216920 W216910 W216911 W216909 W216912 W216908 W216919 W216921 W216913
WHEAW216922EA 30.7 EUR
WHEAW216922 WHEAW216923 215-4448 215-4449 215-4450 215-4451 215-4452 WHEAW216920 215-4455 215-4456 215-4454 215-4457 215-4453 WHEAW216919 WHEAW216921 WHEAW216913
WHEATON® Jars, Wide Mouth, with Thread
Tarros Tarros para uso general
Jars, made of clear soda-lime glass, straight sided.
  • Wide mouth design is great for large solid samples
  • Walls allow for complete removal of contents
  • Ideal for soil sampling and environmental applications
  • Available with caps attached or bulk packs without caps
Certificaciones: Conforms to USP Type III requirements.