Bottles, Wide Neck, Square, Without Screw Cap
Proveedor: RIXIUS
11301010032OSV 11301025045OSV 11301050054OSV 11301100054OSV
215-4667EA 50.1 EUR
215-4667 215-4668 215-4669 215-4670
Bottles, Wide Neck, Square, Without Screw Cap
Frascos Botellas de vidrio
Soda-lime glass, amber.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
DIN thread. Space saving in storage and transport. Stable, non-slip. Amber coloured to protect light-sensitive materials.
Información para pedidos: Screw caps must be ordered separately.