DURAN® protect+ Laboratory Bottles, Plastic Safety Coated, DIN 168-1 Thread, Graduated, Clear
Proveedor: DWK Life Sciences
218051781 218056385 218057381 218051481 218052486 218053688 218055483 218054487 218056985
215-4855EA 22.5 EUR
215-4855 215-4860 215-4862 215-4854 215-4856 215-4857 215-4859 215-4858 215-4861
DURAN® protect+ Laboratory Bottles, Plastic Safety Coated, DIN 168-1 Thread, Graduated, Clear
Frascos Botellas de vidrio
Multi-functional glass laboratory bottles with protective safety coating.
  • Manufactured from Polyamide-coated DURAN® borosilicate glass 3.3
  • Reduce the hazards of broken glass and liquid splashes in the event of breakage
  • External polymer coating provides scratch, leak*, and splinter* protection (*only applies to bottles 2000ml and less)
  • Polyamide 12 safety coating has a high chemical resistance
  • Protective coating provides an enhanced safer grip
  • Coating is cleanroom certified Fraunhofer IPA TESTED DEVICE®
  • Certified FDA and EU food contact safe coating