Airflow detection powder, Draft Checker™, SP Bel-Art
Proveedor: Bel-Art Products, a Part of SP
F199390000
215-6525EA 58.5 EUR
215-6525
Airflow detection powder, Draft Checker™, SP Bel-Art
Materiales para contención de derrames
An amorphous ultra-fine silica powder in a handy squeezy bottle.
  • Suspended powder permits visual estimation of airflow
  • Can be used for detecting leaks around windows, doors and ducts
Safety Note: Silica is inert and non-flammable but appropriate precautions should be taken to prevent ingestion or inhalation of the powder.