Frascos de lavado de boca estrecha con código de colores, Nalgene®
Proveedor: Thermo Fisher Scientific
2422-2500 2422-3500 2422-4500 2422-5500
215-8141EA 59.7 EUR
215-8141 215-8142 215-8143 215-8144
Frascos de lavado de boca estrecha con código de colores, Nalgene®
Frascos Frascos de lavado
High-quality squeeze bottles with colour-coded closures for easy identification.
  • Colour-coded top allows coordination of reagents with industry storage codes
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Sputter-free, precision stream
  • Bottle and carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Nalgene® Colour-Coded LDPE wash bottles are made from lab quality low density polyethylene (More Product Information