Plata nitrato 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS

Proveedor: VWR Chemicals
Peligro

21572.188EA 497 EUR
21572.188 21572.235 21572.133 21572.292
Plata nitrato 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS
Plata nitrato
Formula: AgNO₃
Pto. Ebullición: 444 °C (1013 hPa)
Pto. de fusión: 210 °C
Densidad: 4,35 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003414
Núm. CAS: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
IR: 01-2119513705-43
Índice Merck: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Assay 99.8 - 100.5 %
Clarity of solution Passes test ACS
Free acid Passes test ACS
Identification (Ag) Passes test Ph.Eur.
Identification (NO3) Passes test Ph.Eur.
Solution in water Passes test
Appearance of solution Passes test Ph.Eur.
Acidity or alkalinity Passes test Ph.Eur.
Aluminium, lead, copper and bismuth Passes test Ph.Eur.
Foreign salts ≤ 0.3 %
Insolubility in water ≤ 50 ppm
Not precipitated by HCl (as SO4) ≤ 100 ppm
Cl (Chloride) ≤ 5 ppm
SO4 (Sulphate) ≤ 20 ppm
Ca (Calcium) ≤ 10 ppm
Cd (Cadmium) ≤ 1 ppm
Cu (Copper) ≤ 2 ppm
Fe (Iron) ≤ 2 ppm
K (Potassium) ≤ 50 ppm
Mg (Magnesium) ≤ 5 ppm
Mn (Manganese) ≤ 5 ppm
Ni (Nickel) ≤ 5 ppm
Pb (Lead) ≤ 10 ppm
Zn (Zinc) ≤ 1 ppm
Conforms to Reag. Ph.Eur./ACS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR