Stopcocks for VWR® Collection Carboys
Proveedor: VWR Collection
216-1745EA 22.3 EUR
216-1745
Stopcocks for VWR® Collection Carboys
Válvulas de cierre
PP, white/purple.
Pack consists of stopcock, adapter for tube connection and stopper for long storage. Stopcock is designed to be tightly assembled by hand and, therefore, does not require any tools for assembly. Autoclavable.
Resin meets US FDA 21 CFR, USP Class VI