Containers, straight, wide mouth, with screw cap, Corning® Gosselin™
Proveedor: Corning
TP140-05 TP200-14 TP175-06 TP52C-030 TP52C-032 TP53C-014 TP53C-021 TP53C-023 TP53C-030 TP53C-032 TP53C-042 TP53C-052 TP30C-003 TP30C-004 TP30C-011 TP30C-014 TP30C-021 TP30C-030 TP30C-032 TP35C-001 TP35C-004 TP35C-011 TP35C-021 TP35C-030 TP35C-032 TP35C-042 TP35C-052 TP35C-062 TP52C-001 TP52C-003 TP52C-004 TP52C-021 TP52C-022 TP52C-023 TP52C-024
216-0518EA 325 EUR
216-0518 215-1324 GOSSTP175-06 216-2111 216-2113 GOSSTP53C-014 GOSSTP53C-021 GOSSTP53C-023 GOSSTP53C-030 GOSSTP53C-032 GOSSTP53C-042 GOSSTP53C-052 GOSSTP30C-003 GOSSTP30C-004 216-2084 GOSSTP30C-014 GOSSTP30C-021 216-2085 GOSSTP30C-032 216-2086 216-2087 216-2088 216-2089 216-2091 216-2092 216-2093 216-2094 216-2095 216-2096 216-2097 216-2098 216-2099 216-2100 216-2102 216-2104
Containers, straight, wide mouth, with screw cap, Corning® Gosselin™
Recipientes de muestras Contenedores, uso general
PP translucent or PS clear, with PE screw cap. Straight walled vessels for sample collection, storage and transport. Twist closure for quick opening and closing, with leakproof inner lip. PS offers good thermal and chemical resistance. PP offers excellent thermal and chemical resistance.
  • Selection of colour coded closures for easier sample identification
  • Available sterile or packed under aseptic conditions
Precaución: * Los productos de esta página están fuera del alcance de la Directiva de la UE 2019/904
Order Now
ESPECIFICACIONES
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Certificación ambiental RECs, ISO 14001 - Borre RECs, ISO 14001 - Borre RECs, ISO 14001 - Borre