VWR® Euro Containers, 100% Recycled Polypropylene
Proveedor: VWR Collection
216-2997EA 10.7 EUR
216-2997 216-2998 216-2999 216-3000 216-3001
VWR® Euro Containers, 100% Recycled Polypropylene
Cajas Cajas de almacenamiento
This range of euro containers are made entirely from recycled polypropylene and adhere to the guidelines of the 'Der Blaue Engel' ecological label. Suitable for use in industry, handling, and automated transportation, they remain resilient to impacts and temperature variations. They can be stacked and nested without the risk of falling.
  • Solid stackable and palletisable storage bin made of polypropylene
  • Compliant with european standard for easy storage and transportation
  • Easy handling with open handles
  • Manufactured from recycled polypropylene
  • Lightweight