Tarros, con tapa
Proveedor: Brand
62315
216-3185EA 310 EUR
216-3185
Tarros, con tapa
Tarros Tarros para uso general
PS, with LDPE lid.
  • Conical shape
  • Good chemical resistance
  • Lightweight
Ideal for collecting, storing and transporting samples.