Stoppers, rubber, conical, without holes
Proveedor: DEUTSCH NEUMANN
1010147 1010150 1010156 1010160 1010164 1010171 1010179 1010187 1010194 1010150/12/20
217-9416EA 5.1 EUR
217-9416 217-9417 217-9418 217-9419 217-9420 217-9421 217-9422 217-9423 217-9424 217-0823
Stoppers, rubber, conical, without holes
Tapones Tapones de Goma
Conical natural grey rubber stoppers without holes.
  • Excellent sealing and good adhesion
  • Good chemical resistance
  • Convenient pack sizes
  • Temperature resistant from −25 °C to +70 °C (+120 °C in humid atmosphere)
  • Shore hardness: 40 °A
Elástico, flotante, no tóxico.
Certificaciones: DIN 12871