Anti-CD54 Mouse Monoclonal Antibody [clone: 15.2]

Proveedor: Merck Millipore (Oncogene)
217605-100
217605-100EA 492 EUR
217605-100
Anti-CD54 Mouse Monoclonal Antibody [clone: 15.2]
Anticuerpos
Type: Primary
Antigen: CD54 (15.2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR