Trichoderma viride cellulase
Proveedor: Merck Millipore Calbiochem
219466-100 219466-200 219466-50
219466-100EA 269 EUR
219466-100 219466-200 219466-50
Trichoderma viride cellulase
Enzimas
Cellulase is an effective enzyme to isolate intact cells and protoplasts from plant tissues.
  • Hydrolyses β1, 4-glucan linkages in cellulose
CELLULYSIN® cellulase enzyme shows a very high filter paper decomposing activity, as well as, appreciable xylanase and hemicellulase activity.