Blades for safety knives, No. 60099
Proveedor: MARTOR ARGENTAX
60099.70
223-0112EA 11.9 EUR
223-0112
Blades for safety knives, No. 60099
Cuchillas
Trapezoid blade with rounded tips for use with safety knives.
  • Deep-edged, 2-sided ground-edge
  • Double bevel edge-ground
  • 2-times usable blade
Mountable in:
SECUPRO MAXISAFE
SECUPRO MEGASAFE
SECUPRO 625
SECUNORM MULTISET
SECUNORM PROFI25