Tubo, Tygon® 3350
Proveedor: Saint Gobain Life Sciences
T3304-13 T330G-203 T3302-13 T3302-23 T3304-23 T3308-23 T3310-23 T3306-33 T3308-33 T3308-43 T3316-43 T3303-13 T3306-13 T3308-13 T3312-33 T3314-23 T3324-43 T3320-43 T3305-13 T330N-203 T330P-203 T3310-33 T3303-23 T330B-103 T330H-153 T330L-153 T3312-43 T3306-23 T3312-23 T3316-33 T3314-33
228-0403EA 135 EUR
228-0403 228-4334 228-4331 224-2101 228-4332 228-4333 224-2105 224-2106 224-2107 224-2108 224-2110 228-0402 224-2155 224-2156 224-2157 228-1649 228-1654 228-0637 228-1639 228-4335 228-1646 228-0636 228-1638 228-1642 228-1643 228-1645 228-0698 224-2103 228-0697 228-1652 228-1650
Tubo, Tygon® 3350
Tuberías Tubo de transferencia
Silicone, platinum cured, translucent.
  • Biocompatible, ideal for sensitive applications, can be used for foodstuffs
  • Resistant to ozone and UV, autoclavable
Tygon® tubing made from platinum-cured silicone with ultra-smooth surface. High elasticity with good restoring force for constant liquid transfer, even with intermittent pumping action.
Temperature resistance: −80 to +204 °C
Hardness (Shore durometer A): 50
Density: 1,14
Certificaciones: FDA approved, compliant with USP class VI.
Información de suministro: Tubing is double baggedMore Product Information