Sampling knives, disposable, Sterileware®, SP Bel-Art
Proveedor: Bel-Art Products, a Part of SP
H369650000
231-0016EA 697 EUR
231-0016
Sampling knives, disposable, Sterileware®, SP Bel-Art
Cuchillos
PS, white.
  • Ideal for food sampling, e.g. meats, vegetables, seafood and other fibrous and semi solid materials
  • All knives are lot traceable
  • Jagged blade cuts efficiently, opposite edge can spread material
  • Individually packaged and sterilised
Precaución: * Los productos de esta página están fuera del alcance de la Directiva de la UE 2019/904