Anti-CD45RA Mouse Monoclonal Antibody (APC-Cyanine7) [clone: HI100]

Proveedor: Tonbo Biosciences
25-0458-T025 25-0458-T100
25-0458-T025EA 131 EUR
25-0458-T025 25-0458-T100
Anti-CD45RA Mouse Monoclonal Antibody (APC-Cyanine7) [clone: HI100]
Anticuerpos
The HI100 antibody reacts with the human CD45 isoform known as CD45RA, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns. With their broad cell distribution, CD45 isoforms are critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, includeMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno CD45RA CD45RA
Environmentally Preferable
Tipo de anticuerpo Primario Primario
Clonalidad Monoclonal Monoclonal
Conjugado APC (Allophycocyanin)/Cy7® APC (Allophycocyanin)/Cy7®
Clon HI100 HI100
Reactividad Chimpanzee,Human Chimpanzee,Human
Huésped Mouse Mouse
ID del gen 5788 5788
Isotipo IgG2b kappa IgG2b kappa
Citometría de flujo Yes
Temperatura de almacenamiento 2-8°C protected from light

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR