3-Ethoxy-4-methoxybenzaldehyde ≥99%, Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY

Sinónimos: Apremilast Impurity 28

M-Clarity™ quality level = MQ100

252751-25G 252751-5G
252751-25GEA 244 EUR
252751-25G 252751-5G
3-Ethoxy-4-methoxybenzaldehyde ≥99%, Sigma-Aldrich®
3-Ethoxy-4-methoxybenzaldehyde

3-Ethoxy-4-methoxybenzaldehyde is a derivative of vanillin. 3-Ethoxy-4-methoxybenzaldehyde may be used to synthesise Compound of Designation red 10 binding (CDr10b) to investigate its role in neuroinflammatory diseases.

Formula: C₁₀H₁₂O₃
Pto. de fusión: 51,..53 °C (lit.)
Temperatura de almacenaje: Room Temperature
Número MDL: MFCD00010128
Núm. CAS: 1131-52-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2c454a8e65bd06cd919c0e49104f936be6bd23f&vid=a2c454a8e65bd06cd919c0e49104f936be6bd23f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance (Color) White to Tan
Appearance (Form); Powder, Crystals, Crystalline Powder and/or Chunks Conforms to Requirements
Infrared spectrum Conforms to Structure
Purity (GC) ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR