Cresyl violet perchlorate, Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY
Atención

Sinónimos: Cresyl violet perchlorate, laser grade, 5-iminobenzo[a]phenoxazin-9-amine, perchloric acid

MQ100

255246-100MG 255246-500MG
255246-100MGEA 102 EUR
255246-100MG 255246-500MG
Cresyl violet perchlorate, Sigma-Aldrich®
Cresyl violet perchlorate

Cresyl Violet perchlorate is suitable as a laser dye.


Cresyl Violet perchlorate, also known as oxazine 9, is a laser dye. Cresyl violet is a small diaminobenzooxazine with a moderately sized planar conjugated system. Among its many applications, Cresyl Violet can be used to stain for Nissl substance and nuclei in nervous tissue and the identification of Helicobacter. Cresyl Violet perchlorate has been used to make cresyl violet perchlorate-polymer composite thin films. It also has been used as a fluorescence standard.

Formula: C₁₆H₁₂ClN₃O₅
MW: 361.738 g/mol
Temperatura de almacenaje: Room Temperature
Número MDL: MFCD00012671
Núm. CAS: 41830-80-2
EINECS: 255-561-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3af90c37fdba26ed0b3c77f243812fbe44a085c&vid=a3af90c37fdba26ed0b3c77f243812fbe44a085c&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance (Colour) Dark Green
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wave Length In Ethanol 599 - 605 nm
Extinction Coefficient ≥27000
Wave Length ≥267 - 273 nm
Carbon ≥51.3%
Nitrogen ≥11.2%
Purity (HPLC) ≥96.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR