Sodio fluoruro 99.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent

Proveedor: VWR Chemicals
Peligro

27860.231EA 55.8 EUR
27860.231 27860.297 27860.460
Sodio fluoruro 99.0-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Sodio fluoruro
Formula: NaF
Pto. Ebullición: 1700 °C (1013 hPa)
Pto. de fusión: 993 °C
Densidad: 2,8 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003524
Núm. CAS: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1,III
Índice Merck: 13,08691
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Assay (calculated on dried substance) 99.0 - 100.5 %
Appearance of solution Passes test Ph.Eur.
Identification A Passes test Ph.Eur.
Identification B Passes test Ph.Eur.
Identification C Passes test Ph.Eur.
Acidity ≤ 0.030 meq/g
Alkalinity ≤ 0.010 meq/g
Heavy metals (as Pb) ≤ 10 ppm
Insolubility in water ≤ 100 ppm
Loss on drying (150°C) ≤ 0.2 %
Sodium fluosilicate (Na2SiF6) ≤ 0.1 %
Cl (Chloride) ≤ 100 ppm
SO3 (Sulphite) ≤ 50 ppm
SO4 (Sulphate) ≤ 100 ppm
Fe (Iron) ≤ 20 ppm
K (Potassium) ≤ 0.02 %
Conforms to Reag. Ph.Eur Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR