Sample concentrators, Vivaspin®
Proveedor: Cytiva
28-9322-40 28-9322-37 28-9322-36 28-9322-35 28-9322-25 28-9322-23 28-9322-18 28-9322-47 28-9322-48 28-9323-61 28-9322-45 28-9323-62 28-9323-60 28-9323-59 28-9323-58 28-9323-19 28-9323-18 28-9323-17 28-9322-96 28-9322-94 28-9322-93 28-9322-58 28-9322-57 28-9323-63
28-9322-40EA 216 EUR
28-9322-40 28-9322-37 28-9322-36 28-9322-35 28-9322-25 28-9322-23 28-9322-18 28-9322-47 28-9322-48 28-9323-61 28-9322-45 28-9323-62 28-9323-60 28-9323-59 28-9323-58 28-9323-19 28-9323-18 28-9323-17 28-9322-96 28-9322-94 28-9322-93 28-9322-58 28-9322-57 28-9323-63
Sample concentrators, Vivaspin®
Filtros Filtros centrífugos
Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration.
  • One-step sample concentration in a single tube for minimal sample handling and reduced sample loss
  • Patented dead-stop technology, which ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery
  • Vertical PES membrane minimises membrane blockage and tolerates high flow rates
  • Compatible pH range is from pH 1 to 9
Up to 30-fold concentration of the sample can be achieved with recovery ofMore Product Information
Order Now
ESPECIFICACIONES
Tipo Vivaspin® 2 Vivaspin® 500 Vivaspin® 20 Vivaspin® 6
Área de filtración 1,2 cm² 0,5 cm² 6,0 cm² 2,5 cm²
Environmentally Preferable
Volumen muerto <10 µl <5 µl 50 µl <10 µl
Ø×H 17×126 mm 11×50 mm 30×116 mm 16×122 mm