Protein G Mag Sepharose Xtra, Cytiva
Proveedor: Cytiva
28-9670-66 28-9670-70
28-9670-66EA 124.81 EUR
28-9670-66 28-9670-70
Protein G Mag Sepharose Xtra, Cytiva
Perlas Bolas magnéticas
Protein G Mag Sepharose Xtra magnetic beads are designed for rapid, small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants.
  • Efficient small-scale purification and screening of antibodies from various species; 27 μg human IgG/μl magnetic beads
  • High-capacity, > 27 μg human IgG/μL medium, allows for the efficient capture of the antibody with high-yield and purity Approximately 350 μg purified human IgG can be obtained from each purification run
  • Robust parallel screening of antibodies with high