Anti-IgG Goat Antibody (CyDye™ 700)
Proveedor: Cytiva
29-3607-84 29-3607-85
29-3607-84EA 123.98 EUR
29-3607-84 29-3607-85
Anti-IgG Goat Antibody (CyDye™ 700)
Anticuerpos
Amersham CyDye NIR-labeled secondary antibodies enable quantitation of Western blots with a strong signal-to-noise ratio. Mulitplexed experiments allow for the detection of two proteins on a single blot. The use of total lane signals to normalize target protein data yield robust and reliable results.
  • Easy handling and a strong signal-to-noise ratio yield reproducible results
  • Multiplexing analysis of more than one target protein on a single blot increases process efficiency
  • Total protein normalization (TPN) generates reliable and accurate quantitation