Anti-CD4 Rat Monoclonal Antibody (Biotin) [clone: GK1.5]
Proveedor: Tonbo Biosciences
30-0041-U025 30-0041-U100
30-0041-U025EA 23.5 EUR
30-0041-U025 30-0041-U100
Anti-CD4 Rat Monoclonal Antibody (Biotin) [clone: GK1.5]
Anticuerpos
The GK1.5 antibody reacts with mouse CD4, a 55 kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta-2 domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4 is typically expressed on thymocytes, certain mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells. The GK1.5 antibody is widelyMore Product Information