Disposable samplers, ViscoThief, SteriWare®
Proveedor: Sampling Systems
8055H-1000 8055H-1000S 8055H-500 8055H-500S 8055P-1000 8055P-500 8055H-1500 8055H-1500S
300-2010EA 387 EUR
300-2010 300-2011 300-2012 300-2013 300-2014 300-2015 300-0342 300-0343
Disposable samplers, ViscoThief, SteriWare®
Muestreadores Muestreadores desechables de solo uso
Disposable viscous sampler is designed for creams, pastes and gels. The robust HDPE and PP construction allows for a huge number of liquid and low viscosity testing applications.
  • No risk of cross-contamination (device can be used straight from the bag)
  • Silicone seal for improved sampling performance
  • Assembled and packed in a class 100000 cleanroom
Suitable for taking fast samples from tanks, kegs and containers in industries such as pharmaceutical, food, chemical, cosmetic, biotechnology. Pre-sterilised versions are available.
Certificaciones: FDA,More Product Information