Potasio cloruro 99.0-105.0% (base secada), en cristales USP, Multi-Compendial, J.T.Baker®

Proveedor: Avantor
Atención


Management of Change (MOC) category = R

3045-01EA 89.04 EUR
3045-01 3045-05 3045-07 3045-09
Potasio cloruro 99.0-105.0% (base secada), en cristales USP, Multi-Compendial, J.T.Baker®
Potasio cloruro

Pyrogen tested.


  • GMP manufactured product

Formula: KCl
Pto. Ebullición: 1420 °C (1013 hPa)
Pto. de fusión: 773 °C
Densidad: 1,98 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011360
Núm. CAS: 7447-40-7
EINECS: 231-211-8
Índice Merck: 14,07621

Order Now

Resultados test especificaciones

GMP Manufactured Product 99.0 - 100.5%
Meets J.P. Chemical Specifications Passes Test
Meets U.S.P Requirements ≤0.1%
Meets E.P. Chemical Specifications Passes Test
Meets B.P. Chemical Specifications ≤2.5
Preserve in well-closed containers. Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical ≤0.005%
USP - Assay (KCl) (dried basis) 99.0 - 100.5 %
USP - Acidity or Alkalinity Passes Test
USP - Bromide (Br) ≤ 0.1 %
USP - Calcium and Magnesium Passes Test
USP - Endotoxin Concentration (EU/g) ≤ 2.5
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Iodides (I) ≤ 0.005 %
USP - Loss on Drying ≤ 1.0 %
USP - Sodium (Na) Passes Test
Solution Test Passes Test
EP/BP - Assay (KCl) (dried basis) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Bromide (Br) ≤ 0.1 %
EP/BP - Iodide (I) Passes Test
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Iron (Fe) ≤ 20 ppm
EP/BP - Magnesium and Alkaline Earth Metals ≤ 200 ppm
EP/BP - Loss on Drying at 105°C ≤ 1.0 %
JP - Assay (KCl) (dried basis) 99 - 101 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Acidity or Alkalinity Passes Test
JP - Bromide (Br) Passes Test
JP - Iodide (I) Passes Test
JP - Heavy Metals (as Pb) ≤ 5 ppm
JP - Calcium and Magnesium Passes Test
JP - Sodium (Na) Passes Test
JP - Arsenic (As) ≤ 2 ppm
JP - Loss on Drying ≤ 0.5 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR