Microcentrifuge tubes with attached caps, in resealable bags, SuperClear®
Proveedor: LABCON
3022-800-000-9 3034-335-000-9 3046-870-000-9 3046-871-000-9 3046-872-000-9 3046-873-000-9 3046-874-000-9 3046-876-000-9 3046-877-000-9 3046-878-000-9 3046-879-000-9
3022-800-000-9EA 102 EUR
3022-800-000-9 3034-335-000-9 3046-870-000-9 3046-871-000-9 3046-872-000-9 3046-873-000-9 3046-874-000-9 3046-876-000-9 3046-877-000-9 3046-878-000-9 3046-879-000-9
Microcentrifuge tubes with attached caps, in resealable bags, SuperClear®
Tubos Tubos de microcentrífugas
Ultra-clear polypropylene, graduated, autoclavable, and freezable microcentrifuge tubes with flat cap. Amber coloured styles are available for use with light-sensitive substances.
  • Every lot is tested for protease, Human DNA, ATP, and RNase/DNase
  • Endotoxin-free (non-pyrogenic)
  • Supplied with flat, easy-open, attached snap caps
  • Side graduations and frosted writing surface
  • Polypropylene USP Class VI resin
  • Freezable to –80 °C
The ability to extract DNA from samples and then amplify that DNA with reliable results is an important part of lab work. SuperClear®More Product Information