Monensin methyl ester ≥97.0% (by TLC), Selectophore™ ionophore, Supelco®

Proveedor: Merck
Peligro


M-Clarity™ quality level = MQ100

30552-100MG
30552-100MGEA 194 EUR
30552-100MG
Monensin methyl ester ≥97.0% (by TLC), Selectophore™ ionophore, Supelco®
Monensin methyl ester

Monensin, is an antibiotic produced by Streptomyces cinnamonensis. It is a naturally occurring ionophore.


It has an acyclic polyether sequence and a terminal carboxylic acid moiety. It has the ability to effectively carry protons and cations through the cell membrane, hence it adequately harbours Na+ into a characteristic pseudoactivity, thereby effectively transporting it across a bio-membrane. It also forms stable complexes with k+, Rb+ and Ag+; hence it can be chemically modified to form cation-binding sites, thereby attaining transport functionalities.

Formula: C₃₇H₆₄O₁₁
Núm. CAS: 28636-21-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe85da9ff1b9f4e3a35cf30793ff95a8acd60e8c&vid=910922959098e368ca145cab5929f16053586004&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR