Sodio hidróxido ≥99.99% (trace metals basis), en lentejas, Semiconductor grade

Proveedor: Honeywell Chemicals
Peligro

306576-100G 306576-25G 306576-500G
306576-100GEA 186 EUR
306576-100G 306576-25G 306576-500G
Sodio hidróxido ≥99.99% (trace metals basis), en lentejas, Semiconductor grade
Sodio hidróxido

  • Comprehensive portfolio for the electronics and semiconductor industry
  • For scientists engaged in education, research, development, prototyping and small scale fabrication of high tech micro- and nanochips
  • Superior purity, stability and exceptional lot-to-lot consistency
  • Includes high purity products required for high performance applications such as etchants, solvents or hexamethyldisilazane (HMDS)

Formula: NaOH
Pto. Ebullición: 1,390 °C (1.013 hPa)
Pto. de fusión: 319 °C
Densidad: 2,13 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003548
Núm. CAS: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
IR: 01-2119457892-27
Índice Merck: 14,08627

Order Now

Resultados test especificaciones

Vapor Pressure 3 mmHg ( 37 °C)
Appearance of the substance complying
Assay Min. 95.0 %
Assay of Na₂CO₃ Max. 1.0 %
Boron (B) Max. 10 ppm
Cesium (Cs) Max. 20 ppm
Potassium (K) Max. 100 ppm
Lithium (Li) Max. 10 ppm
Rubidium (Rb) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR