Insulating tubes, ceramic
Proveedor: Thermo Fisher Scientific
31475.DR 31476.DR 31476.DT 31479.DR 31479.DW 31492.DR 31475.DT 31475.DW 31485.DR 31485.DW 31487.DR 31487.DW
31475.DREA 36.1 EUR
31475.DR 31476.DR 31476.DT 31479.DR 31479.DW 31492.DR 31475.DT 31475.DW 31485.DR 31485.DW 31487.DR 31487.DW
Insulating tubes, ceramic
Tuberías Tubo de transferencia
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.