Cloroformo ≥99.8% estabilizado ACS, Riedel-de Haën™

Proveedor: Honeywell Chemicals
Peligro

Sinónimos: Triclorometano

Estabilización: estabilizado con etanol 0,5-1,0%

319988-500ML
319988-500MLEA 68 EUR
319988-500ML
Cloroformo ≥99.8% estabilizado ACS, Riedel-de Haën™
Cloroformo

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications

Formula: CHCl₃
Pto. Ebullición: 60…62 °C (1,013 hPa)
Pto. de fusión: –63 °C
Densidad: 1,48 g/cm3 (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00000826
Núm. CAS: 67-66-3
UN: 1888
ADR: 6.1,III
IR: 05-2114614906-45
Índice Merck: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Vapor Pressure 160 mmHg ( 20 °C)
Identity (IR) complying
Purity excluding stabilizer (GC) Min. 99.8 %
Ethanol (GC) 0.5 - 1.0 Gew.%
Non-volatile matter Max. 0.001 %
Lead (Pb) Max. 0.05 ppm
Free chlorine complying
Free acid and chloride complying
Acetone, aldehydes (neßler test) complying
Reaction against H₂SO₄ complying
APHA Max. 10
Vapor Density 4.1 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR