Acetona ≥99.5% (por GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™

Proveedor: Honeywell Chemicals
Peligro

32201-2.5L-GL 32201-7L 32201-1L 32201-2.5L 32201-20KG 32201-25L 32201-4X2.5L 32201-6X1L 32201-4X2.5LGL 32201-18L 32201-4X2.5L-G
32201-2.5L-GLEA 117 EUR
32201-2.5L-GL 32201-7L 32201-1L 32201-2.5L 32201-20KG 32201-25L 32201-4X2.5L 32201-6X1L 32201-4X2.5L-GL HONC32201-18L HONC32201-4X2.5L-G
Acetona ≥99.5% (por GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Acetona

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 635680
RTECS Number: AL3150000
UNSPSC Code: 12191602

Formula: CH₃COCH₃
Pto. Ebullición: 55…57 °C (1,013 hPa)
Pto. de fusión: –95 °C
Densidad: 0,79 g/cm³ (20 °C)
Pto. de inflamación: 14,3 %(V)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00008765
Núm. CAS: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
IR: 01-2119471330-49
Índice Merck: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Assay (GC) Min. 99.5 %
Boiling range 55.5 - 56.5 °C
Density (D 20/4) 0.790 - 0.792
Refractive index (n 20/D) 1.3580 - 1.3600
Non-volatile matter Max. 10 ppm
Water insoluble substances complying
Water (Karl Fischer) Max. 0.2 %
Free acid (as CH₃COOH) Max. 0.0015 %
Free alkali (as NH₃) Max. 0.0008 %
Acidity or alkalinity complying
Aluminium (Al) Max. 0.5 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.02 ppm
Tin (Sn) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.05 ppm
KMnO₄ reducing matter (as O) Max. 0.0002 %
Reducing impurities complying
Aldehydes (as HCHO) Max. 0.001 %
Benzene (GC) Max. 2 Vol.ppm
Ethanol (GC) Max. 0.01 %
Methanol (GC) Max. 0.05 Vol.%
2-propanol (GC) Max. 0.05 Vol.%
Related substances (GC) complying
Appearance of the solution complying
APHA Max. 10
Solubility in water complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR