Ciclohexano ≥99.5% (por GC), Puriss. p.a. ACS, Riedel-de Haën™

Proveedor: Honeywell Chemicals
Peligro

33117-18L 33117-1L 33117-2.5L 33117-30KG 33117-4X2.5L 33117-5L 33117-6X1L 33117-30L-RC 33117-200L 33117-45L-RC 33117S-7L-RC
33117-18LEA 1630 EUR
33117-18L 33117-1L 33117-2.5L 33117-30KG 33117-4X2.5L 33117-5L 33117-6X1L 33117-30L-RC HONC33117-200L HONC33117-45L-RC HONC33117S-7L-RC
Ciclohexano ≥99.5% (por GC), Puriss. p.a. ACS, Riedel-de Haën™
Ciclohexano

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 1900225
RTECS Number: GU6300000
UNSPSC Code: 12191504

Formula: C₆H₁₂
Pto. Ebullición: 81 °C (1,013 hPa)
Pto. de fusión: 6,4 °C
Densidad: 0,78 g/cm³ (20 °C)
Pto. de inflamación: 8,3 %(V)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003814
Núm. CAS: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Índice Merck: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance complying
Assay (GC) Min. 99.5 %
Solidification range 5.5 - 6.5 °C
Boiling range 79 - 81 °C
Vapor Density 2.9 (vs air)
Transition Temp solidification point 5.5-6.5 °C
Density (D 20/4) 0.778 - 0.779
Refractive index (n 20/D) 1.4250 - 1.4280
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as HCl) Max. 0.001 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Aromatic substances (as C₆H₆) Max. 0.05 %
cyclohexene (C₆H₁₀) Max. 0.05 %
Reaction against H₂SO₄ complying
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR