Etilo acetato ≥99.5% (por GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™

Proveedor: Honeywell Chemicals
Peligro

33211-7L 33211-18L 33211-45L 33211-180KG 33211-1L 33211-2.5L 33211-25L 33211-4X2.5L 33211-6X1L 33211-1000L
33211-7LEA 442 EUR
33211-7L 33211-18L-R 33211-45L-R 33211-180KG 33211-1L 33211-2.5L 33211-25L 33211-4X2.5L 33211-6X1L HONC33211-1000L
Etilo acetato ≥99.5% (por GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Etilo acetato

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 506104
RTECS Number: AH5425000
UNSPSC Code: 12191602

Formula: CH₃COOC₂H₅
Pto. Ebullición: 77 °C (1,013 hPa)
Pto. de fusión: –84 °C
Densidad: 0,901 g/cm³
Pto. de inflamación: 11,5 %(V)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00009171
Núm. CAS: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
IR: 01-2119475103-46
Índice Merck: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Assay (GC) Min. 99.5 %
Boiling range 76 - 78 °C
Density (D 20/4) 0.899 - 0.901
Refractive index (n 20/D) 1.3710 - 1.3730
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as CH₃COOH) Max. 0.0045 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Potassium (K) Max. 0.00005 %
Lithium (Li) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Molybdenum (Mo) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Strontium (Sr) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
methyl acetate Max. 0.1 %
Ethanol (GC) Max. 0.1 %
Isobutyl alcohol (GC) Max. 0.1 %
Methanol (GC) Max. 0.1 %
Reaction against H₂SO₄ complying
APHA Max. 10

ESPECIFICACIONES

Environmentally Preferable Safer Human and Environmental Health Safer Human and Environmental Health Safer Human and Environmental Health
Certificación ambiental Chem21 Solvent Selection Guide recommended by default and after discussion Chem21 Solvent Selection Guide recommended by default and after discussion Chem21 Solvent Selection Guide recommended by default and after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR