Sodio piruvato 99,0 Non aqueous titration, powder_white, BAKER ANALYZED®, J.T.Baker®

Proveedor: Avantor

Sinónimos: Piruvato de sodio

3354-02EA 1000.51 EUR
3354-02 3354-04
Sodio piruvato 99,0 Non aqueous titration, powder_white, BAKER ANALYZED®, J.T.Baker®
Sodio piruvato

Lot analysis on label.

Formula: CH₃COCOONa
MW: 110.04 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00002586
Núm. CAS: 113-24-6
EINECS: 204-024-4

Order Now

Resultados test especificaciones

For Laboratory,Research,or Manufacturing Use 1% max.
Assay (CH₃COCOONa) (by non-aqueous titration) ≥99.0 %
Free Pyruvic Acid (CH₃COCOOH) ≤1 %
Solubility Passes Test
Meets Reagent Specifications for testing USP/NF monographs.
Store cold. Keep Dry. Warm to room temperature before opening.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR