Organic reference standard, Aflatoxin B2 solution, analytical standard, Supelco®
Proveedor: Merck
34034-2ML-R
34034-2ML-REA 293 EUR
34034-2ML-R
Organic reference standard, Aflatoxin B2 solution, analytical standard, Supelco®
Normativas Estándares orgánicos
Aflatoxins are mycotoxins that are generated as a result of secondary metabolism of various fungal species such as AspergillusPenicillium and Fusarium.
Aflatoxin B₂ may be used for the determination of aflatoxin B₂ in: Animal feed by high performance liquid chromatography with fluorescence detection (HPLC-FLD) as well as quick, easy, cheap, effective, rugged and safe (QuEchERS) extraction method combined with ultra-high performance liquid chromatography-high resolution mass spectrometry (UHPLC-HRMS). Eggs by QuEchERS extraction method followed by analysis using UHPLC with tandem mass spectrometry (MS/MS) operating on electrospray-ionization interface (ESI) mode. Brazil nuts by immunoaffinity column (IAC) and clean-up HPLC-FLD. Plant-based milk and dairy products by dispersive liquid-liquid microextraction (DLLME), QuEchERS extraction method and HPLC-FLD.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cb100dad1387a6901f57a781e57ab1e4eaf6592&vid=4cb100dad1387a6901f57a781e57ab1e4eaf6592&prid=web&clid=SAPDAM