1,9-Dimethyl-Methylene Blue zinc chloride double salt 80% Dye, Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY

Sinónimos: DMMB, Taylor’s Blue

341088-1G
341088-1GEA 169 EUR
341088-1G
1,9-Dimethyl-Methylene Blue zinc chloride double salt 80% Dye, Sigma-Aldrich®
1,9-Dimethyl-Methylene Blue zinc chloride double salt
Formula: C₁₈H₂₂ClN₃S·0.5ZnCl₂
MW: 416.05 g/mol
Número MDL: MFCD19686959
Núm. CAS: 931418-92-7

Order Now

Resultados test especificaciones

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
ICP Major Analysis, Confirms Zinc Component Confirmed
Wavelength 243 - 249 nm
Extinction Coefficient ≥25000
Extinction Coefficient ≥18000
Extinction Coefficient, c = 0.003 g/L; Water ≥10000
Carbon ≥40.5%
Nitrogen ≥7.9%
Water (by Karl Fischer) ≤20.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR