FluorSave™ reagent, Calbiochem®
Proveedor: Merck Millipore Calbiochem
345789-20
345789-20EA 125 EUR
345789-20
FluorSave™ reagent, Calbiochem®
Sistemas de enlace y etiquetado de proteínas
FluorSave™ reagent is an aqueous mounting medium for use in conjunction with fluorescent tracers such as fluorescein, rhodamine and phycobiliprotein probes.
  • Preserves fluorescence-stained tissue sections
  • Formulated without glycerol
  • Ideal for use with phycobiliprotein tracers
Order Now
ESPECIFICACIONES
Environmentally Preferable
Forma Liquid
Formulación Formulated without glycerol
Huésped
Temperatura de almacenamiento 15 - 30 °C