Anti-CD45RO Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: UCHL1]
Proveedor: Tonbo Biosciences
35-0457-T100 35-0457-T025
35-0457-T100EA 68.2 EUR
35-0457-T100 35-0457-T025
Anti-CD45RO Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: UCHL1]
Anticuerpos
The UCHL1 antibody reacts with the human CD45 isoform known as CD45RO, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns. With their broad cell distribution, CD45 isoforms are critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, includeMore Product Information