Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct

Proveedor: Thermo Fisher Scientific
Atención

35445.02 35445.04
35445.02EA 47.1 EUR
35445.02 35445.04
Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct
Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct

CAS No.: 75965-35-4 Molecular Formula: C24H20BLi.3C4H10O2 Formula Weight: 595.54 (326.18anhy) Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: White to off-white UN No.: UN1325 MDL No.: MFCD00013311

Formula: C₂₄H₂₀BLi.3C₄H₁₀O₂
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00013311
Núm. CAS: 75965-35-4
ADR: 4.1,III

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR