Screw caps for SuperClear® microtubes
Proveedor: LABCON
3600-870-000-9 3600-871-000-9 3600-872-000-9 3600-873-000-9 3600-874-000-9 3600-876-000-9 3600-877-000-9 3600-878-000-9 3600-879-000-9 3602-870-000-9 3602-871-000-9 3602-872-000-9 3602-873-000-9 3602-874-000-9 3602-876-000-9 3602-877-000-9 3602-878-000-9 3602-879-000-9
3600-870-000-9EA 133 EUR
3600-870-000-9 3600-871-000-9 3600-872-000-9 3600-873-000-9 3600-874-000-9 3600-876-000-9 3600-877-000-9 3600-878-000-9 3600-879-000-9 3602-870-000-9 3602-871-000-9 3602-872-000-9 3602-873-000-9 3602-874-000-9 3602-876-000-9 3602-877-000-9 3602-878-000-9 3602-879-000-9
Screw caps for SuperClear® microtubes
Tapas Tapones para tubos de ensayo
Caps only.
  • Endotoxin Free (non-pyrogenic)
  • Polyethylene USP Class VI resin
  • Polypropylene
  • Autoclavable