Zirconium disilicide ≥99.5% (metals basis excluding Hf) (max. 4% Hf)

Proveedor: Thermo Fisher Scientific

36322.14 36322.22
36322.14EA 138 EUR
36322.14 36322.22
Zirconium disilicide ≥99.5% (metals basis excluding Hf) (max. 4% Hf)
Zirconium disilicide

Purity: 99.5% CAS No.: 12039-90-6 Molecular Formula: ZrSi2 (may contain other phases) Formula Weight: 147.39 Physical Form: -325 Mesh Powder Density: 4.88 Appearance: Grey Odour: Odourless MDL No.: MFCD00151547

Formula: Si₂Zr
Densidad: 4,88
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00151547
Núm. CAS: 12039-90-6
EINECS: 234-911-1

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR