Sodio dihidrogenofosfato monohidrato 98,0-103,0 Anhydrous basis, crystals USP, FCC Endotoxins_tested, J.T.Baker®

Proveedor: Avantor

Sinónimos: Sodio fosfato prim. monohidrato

3821-07EA 1179.64 EUR
3821-07 3821-01
Sodio dihidrogenofosfato monohidrato 98,0-103,0 Anhydrous basis, crystals USP, FCC Endotoxins_tested, J.T.Baker®
Sodio dihidrogenofosfato monohidrato

  • Food GMP manufactured product

Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Pto. de fusión: 100
Densidad: 2.04 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00149208
Núm. CAS: 10049-21-5
EINECS: 231-449-2
IR: 01-2119489796-13
Índice Merck: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Meets F.C.C. Requirements 98.0 - 103.0%
Meets U.S.P Requirements Passes Test
GMP Manufactured Product ≤3 ppm
Food GMP Manufactured Product ≤0.014%
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Food Chemical Passes Test
Bulk Pharmaceutical Chemical ≤0.2%
USP - Assay (anhydrous basis) 98.0 - 103.0 %
USP - Aluminum, Calcium, and Related Elements Passes Test
USP - Arsenic (As) ≤3 ppm
USP - Chloride (Cl) ≤0.014 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Substances ≤0.2 %
USP - pH (1 in 20) 4.1 - 4.5
USP - Sulfate (SO₄) ≤0.15 %
USP - Water (H₂O) 10.0 - 15.0 %
FCC - Assay (anhydrous basis) 98.0 - 103.0 %
FCC - Arsenic (As) ≤3 ppm
FCC - Fluoride (F) ≤0.005 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Insoluble Substances ≤0.2 %
FCC - Lead (Pb) ≤4 mg/kg
FCC - Loss on Drying 10.0 - 15.0 %
Endotoxin Concentration (EU/g) ≤2.5
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR