Sulfur in diesel fuel standard solutions, Specpure®

Proveedor: Thermo Fisher Scientific

Sulfur in diesel fuel standard solutions, Specpure®
Normativas Estándares de la industria petrolera

Specpure® calibration standards, designed specifically for the analysis of sulphur in petroleum products by X-ray fluorescence spectrometry.


Physical Form: Liquid
MDL No.: MFCD01095233

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR