Dishes, aluminium, flat bottom, straight wall
Proveedor: Thermo Fisher Scientific
39072.KH 39072.UD
39072.KHEA 43.4 EUR
39072.KH 39072.UD
Dishes, aluminium, flat bottom, straight wall
Bandejas/Platos/Ollas Bandejas/Placas de laboratorio
Dishes, Aluminium, flat bottom, straight sides.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Aluminum is the most abundant metallic element on the earth and has many desirable physical and chemical properties. A highly impervious oxide film (approximately 5nm-thick) is resistant to corrosion by seawater, and other aqueous and chemical solutions. Additionally, this element offers chemical stability in the presence of most organic compounds. Aluminum's reactivity increases with temperature, therefore,More Product Information