Sealing film, self-adhesive for PCR, real time PCR
Proveedor: Brand
781383
391-0705EA 98.2 EUR
391-0705
Sealing film, self-adhesive for PCR, real time PCR
Juntas Sellos de microplaca
Polyester flims suitable for PCR and Real-Time PCR measurements. Temperature range: –40 °C to +120 °C.