Micro-cavity plates, Corning® Elplasia®
Proveedor: Corning
4440 4441 4442 4444 4445 4446 4447
CORN4440EA 593.82 EUR
CORN4440 CORN4441 391-1006 CORN4444 CORN4445 CORN4446 CORN4447
Micro-cavity plates, Corning® Elplasia®
Microplacas Placas de cultivo celular
Elplasia plates enable researchers to generate high density of spheroids in a scaffold-free model. Use Corning Elplasia plates to generate, culture and analyse your spheroids all in a standard plate footprint.
  • Black opaque side walls reduce well-to-well cross-talk
  • Gamma irradiated
  • Suitable for fluorescent/luminescent assays
  • One year shelf life
These plates are compatible with many cell types and may be used across many applications including: Drug/high throughput screening, cancer/tumour biology, stem cell biology, cell therapy research and 3-D tissueMore Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable Low Manufacturing Impact
Certificación ambiental RECs, ISO 14001 - Oneonta